MAT196009421 Matrix Scientific

[(4-Fluorobenzyl)sulfonyl]acetic acid

  • MDL number MFCD06208212
  • Cas Number : 222639-41-0
  • Molecular Weight : 232.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040419.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040419.html
Availability details for
Quantity
  • Availability
  • Add to Cart