MAT196009428 Matrix Scientific

2-(Isopropylthio)benzoic acid

  • MDL number MFCD07364761
  • Cas Number : 41394-95-0
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040426.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040426.html
Availability details for
Quantity
  • Availability
  • Add to Cart