MAT196009429 Matrix Scientific

2-(Isopropylsulfonyl)benzoic acid

  • MDL number MFCD09934659
  • Cas Number : 20884-63-3
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040427.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040427.html
Availability details for
Quantity
  • Availability
  • Add to Cart