MAT196009432 Matrix Scientific

[(3,4-Dichlorobenzyl)sulfonyl]acetic acid

  • MDL number MFCD10691573
  • Cas Number : 300700-03-2
  • Molecular Weight : 283.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040430.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040430.html
Availability details for
Quantity
  • Availability
  • Add to Cart