MAT196009654 Matrix Scientific

(3,5-Dimethyl-1H-pyrazol-1-yl)acetaldehyde

  • MDL number MFCD04969649
  • Cas Number : 894694-12-3
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040235.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040235.html
Availability details for
Quantity
  • Availability
  • Add to Cart