MAT196009729 Matrix Scientific

1-(Methylsulfonyl)-4-nitro-1H-pyrazole

  • MDL number MFCD04969872
  • Cas Number : 1005640-95-8
  • Molecular Weight : 191.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040727.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040727.html
Availability details for
Quantity
  • Availability
  • Add to Cart