MAT196009740 Matrix Scientific

2-(2-Chlorophenyl)azepane

  • MDL number MFCD07357623
  • Cas Number : 887360-60-3
  • Molecular Weight : 209.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040738.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040738.html
Availability details for
Quantity
  • Availability
  • Add to Cart