MAT196009743 Matrix Scientific

1-(Pyrrolidin-2-ylmethyl)-1H-pyrazole

  • MDL number MFCD09859232
  • Cas Number : 1171334-97-6
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040741.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040741.html
Availability details for
Quantity
  • Availability
  • Add to Cart