MAT196009752 Matrix Scientific

(3-Isopropyl-4,5-dihydroisoxazol-5-yl)methanol

  • MDL number MFCD12028399
  • Cas Number : 1142210-97-6
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040750.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040750.html
Availability details for
Quantity
  • Availability
  • Add to Cart