MAT196009765 Matrix Scientific

Methyl 4-(1-ethyl-1H-pyrazol-4-yl)-2,4-dioxobutanoate

  • MDL number MFCD06739356
  • Cas Number : 1006483-29-9
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040763.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040763.html
Availability details for
Quantity
  • Availability
  • Add to Cart