MAT196009777 Matrix Scientific

[3-(4-Chlorophenyl)-5-isoxazolyl]methanamine

  • MDL number MFCD01935956
  • Cas Number : 66046-42-2
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/040775.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040775.html
Availability details for
Quantity
  • Availability
  • Add to Cart