MAT196009795 Matrix Scientific

6-Bromo-3-chloropyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD00297265
  • Cas Number : 499190-16-8
  • Molecular Weight : 276.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040793.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040793.html
Availability details for
Quantity
  • Availability
  • Add to Cart