MAT196009820 Matrix Scientific

5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo-[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD02253965
  • Cas Number : 524036-15-5
  • Molecular Weight : 337.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040818.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040818.html
Availability details for
Quantity
  • Availability
  • Add to Cart