MAT196009871 Matrix Scientific

5-(1H-Tetrazol-1-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD12028417
  • Cas Number : 1039008-40-6
  • Molecular Weight : 180.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040869.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040869.html
Availability details for
Quantity
  • Availability
  • Add to Cart