MAT196009881 Matrix Scientific

Ethyl 3-amino-1H-pyrazole-5-carboxylate

  • MDL number MFCD11215301
  • Cas Number : 105434-90-0
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040879.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040879.html
Availability details for
Quantity
  • Availability
  • Add to Cart