MAT196009896 Matrix Scientific

5-Ethylisoxazole-3-carbonyl chloride

  • MDL number MFCD12028420
  • Cas Number : 52320-58-8
  • Molecular Weight : 159.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040894.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040894.html
Availability details for
Quantity
  • Availability
  • Add to Cart