MAT196009898 Matrix Scientific

3,5-Dimethylisoxazole-4-carbonyl chloride

  • MDL number MFCD00052555
  • Cas Number : 31301-45-8
  • Molecular Weight : 159.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040896.html

Safety Information

hcodes Corrosive/Moisture Sensitive
MSDS http://www.matrixscientific.com/040896.html
Availability details for
Quantity
  • Availability
  • Add to Cart