MAT196009907 Matrix Scientific

3-(Butyrylamino)benzoic acid

  • MDL number MFCD00448227
  • Cas Number : 76209-00-2
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040905.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040905.html
Availability details for
Quantity
  • Availability
  • Add to Cart