MAT196009924 Matrix Scientific

3-(4-Hydroxyphenyl)propanohydrazide

  • MDL number MFCD00025134
  • Cas Number : 65330-63-4
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040922.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040922.html
Availability details for
Quantity
  • Availability
  • Add to Cart