MAT196009941 Matrix Scientific

2-(2-Aminoethyl)quinazolin-4(3H)-one hydrochloride

  • MDL number MFCD11046440
  • Cas Number : 1185298-40-1
  • Molecular Weight : 225.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040939.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040939.html
Availability details for
Quantity
  • Availability
  • Add to Cart