MAT196009958 Matrix Scientific

tert-Butyl (3S,4R)-3-(hydroxymethyl)-4-(4-methoxyphenyl)pyrrolidine-1-carboxylate

  • MDL number MFCD12028189
  • Cas Number : 1186654-76-1
  • Molecular Weight : 307.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040315.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040315.html
Availability details for
Quantity
  • Availability
  • Add to Cart