MAT196009963 Matrix Scientific

(3S,4R)-1-(2-Ethoxyethyl)-4-(3-methoxyphenyl)-pyrrolidine-3-carboxylic acid

  • MDL number MFCD12028192
  • Cas Number : 1186648-00-9
  • Molecular Weight : 293.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040319.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040319.html
Availability details for
Quantity
  • Availability
  • Add to Cart