MAT196009982 Matrix Scientific

tert-Butyl 5-oxo-1,4-diazepane-1-carboxylate

  • MDL number MFCD04115319
  • Cas Number : 190900-21-1
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040980.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040980.html
Availability details for
Quantity
  • Availability
  • Add to Cart