MAT196009994 Matrix Scientific

{1-[(tert-Butoxycarbonyl)amino]cyclobutyl}-methyl tert-butyl carbonate

  • MDL number MFCD12028468
  • Cas Number : 1142211-21-9
  • Molecular Weight : 301.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040992.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040992.html
Availability details for
Quantity
  • Availability
  • Add to Cart