MAT196009999 Matrix Scientific

(2,3-Dimethyl-1H-indol-5-yl)methanol

  • MDL number MFCD12028472
  • Cas Number : 1071667-99-6
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040997.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040997.html
Availability details for
Quantity
  • Availability
  • Add to Cart