MAT196010023 Matrix Scientific

5-Bromo-2-furamide

  • MDL number MFCD00020926
  • Cas Number : 6134-61-8
  • Molecular Weight : 190

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041021.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041021.html
Availability details for
Quantity
  • Availability
  • Add to Cart