MAT196010027 Matrix Scientific

3-Fluoro-2-methoxypyridine

  • MDL number MFCD03094945
  • Cas Number : 884494-69-3
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.146
Flash Point 38
Storage temp. Ambient
Country of Origin USA
Boiling point 139
Product Link http://www.matrixscientific.com/041025.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041025.html
Availability details for
Quantity
  • Availability
  • Add to Cart