MAT196010037 Matrix Scientific

tert-Butyl (2-hydroxybenzyl)(2-hydroxyethyl)-carbamate

  • MDL number MFCD12028206
  • Cas Number : 1142202-20-7
  • Molecular Weight : 267.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040345.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040345.html
Availability details for
Quantity
  • Availability
  • Add to Cart