MAT196010045 Matrix Scientific

Ethyl 5-bromo-4-nitrothiophene-2-carboxylate

  • MDL number MFCD12028210
  • Cas Number : 2160-52-3
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040351.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040351.html
Availability details for
Quantity
  • Availability
  • Add to Cart