MAT196010075 Matrix Scientific

5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01203002
  • Cas Number : 15884-86-3
  • Molecular Weight : 145.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040377.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040377.html
Availability details for
Quantity
  • Availability
  • Add to Cart