MAT196010089 Matrix Scientific

4-(5-Amino-1,3,4-thiadiazol-2-yl)-1-phenylpyrrolidin-2-one

  • MDL number MFCD12028219
  • Cas Number : 1142202-31-0
  • Molecular Weight : 260.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040391.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040391.html
Availability details for
Quantity
  • Availability
  • Add to Cart