MAT196010091 Matrix Scientific

4-(5-Amino-1,3,4-thiadiazol-2-yl)-1-(2-methylphenyl)pyrrolidin-2-one

  • MDL number MFCD12028221
  • Cas Number : 1142202-37-6
  • Molecular Weight : 274.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040393.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040393.html
Availability details for
Quantity
  • Availability
  • Add to Cart