MAT196010095 Matrix Scientific

4-(5-Amino-1,3,4-thiadiazol-2-yl)-1-(4-fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD12028225
  • Cas Number : 1142202-45-6
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040397.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040397.html
Availability details for
Quantity
  • Availability
  • Add to Cart