MAT234723095 Matrix Scientific

2-(Isopentyloxy)-4-methylaniline hydrochloride

  • MDL number MFCD12405028
  • Cas Number : 1185295-20-8
  • Molecular Weight : 229.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022185.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022185.html
Availability details for
Quantity
  • Availability
  • Add to Cart