MAT234723098 Matrix Scientific

4-(Benzyloxy)-3-methylaniline hydrochloride

  • MDL number MFCD12025952
  • Cas Number : 1150114-24-1
  • Molecular Weight : 249.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022657.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022657.html
Availability details for
Quantity
  • Availability
  • Add to Cart