MAT234723115 Matrix Scientific

4-(1-Methyl-1-phenylethyl)phenyl4-piperidinylether hydrochloride

  • MDL number MFCD12405047
  • Cas Number : 1185296-87-0
  • Molecular Weight : 331.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023303.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023303.html
Availability details for
Quantity
  • Availability
  • Add to Cart