MAT234723119 Matrix Scientific

3-(4-Benzylphenoxy)pyrrolidine hydrochloride

  • MDL number MFCD12405051
  • Cas Number : 1185297-42-0
  • Molecular Weight : 289.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023414.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023414.html
Availability details for
Quantity
  • Availability
  • Add to Cart