MAT234723121 Matrix Scientific

2-[2-(3-Fluorophenoxy)ethyl]piperidinehydrochloride

  • MDL number MFCD12405053
  • Cas Number : 1185299-58-4
  • Molecular Weight : 259.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023451.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023451.html
Availability details for
Quantity
  • Availability
  • Add to Cart