MAT234723126 Matrix Scientific

2-Chloro-4-fluoro-3-methoxybenzyl bromide

  • MDL number MFCD09835511
  • Cas Number : 1171917-24-0
  • Molecular Weight : 253.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041122.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041122.html
Availability details for
Quantity
  • Availability
  • Add to Cart