MAT234723146 Matrix Scientific

2-(N-Morpholinyl)-6-fluorobenzaldehyde

  • MDL number MFCD11855806
  • Cas Number : 736991-93-8
  • Molecular Weight : 209.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041146.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041146.html
Availability details for
Quantity
  • Availability
  • Add to Cart