MAT234723148 Matrix Scientific

3-Fluoro-4-(N-morpholino)phenol

  • MDL number MFCD11974203
  • Cas Number : 1171917-53-5
  • Molecular Weight : 197.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041148.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041148.html
Availability details for
Quantity
  • Availability
  • Add to Cart