MAT235990778 Matrix Scientific

2-Methyl-4-(phenethyloxy)aniline hydrochloride

  • MDL number MFCD12547704
  • Cas Number : 1185296-12-1
  • Molecular Weight : 263.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-183
Country of Origin USA
Product Link http://www.matrixscientific.com/022806.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022806.html
Availability details for
Quantity
  • Availability
  • Add to Cart