MAT235990782 Matrix Scientific

2-(3-Piperidinyl)ethyl propyl ether hydrochloride

  • MDL number MFCD12405058
  • Cas Number : 946727-29-3
  • Molecular Weight : 207.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023461.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023461.html
Availability details for
Quantity
  • Availability
  • Add to Cart