MAT236031695 Matrix Scientific

Di-Fmoc-3,4-diaminobenzoic acid

  • MDL number MFCD02682246
  • Cas Number : 345958-22-7
  • Molecular Weight : 596.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/041702.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041702.html
Availability details for
Quantity
  • Availability
  • Add to Cart