MAT236031706 Matrix Scientific

Fmoc-3-amino-4-methylbenzoic acid

  • MDL number MFCD01861379
  • Cas Number : 1072901-59-7
  • Molecular Weight : 373.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041727.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041727.html
Availability details for
Quantity
  • Availability
  • Add to Cart