MAT236031748 Matrix Scientific

Fmoc-L-selenomethionine

  • MDL number MFCD03093662
  • Cas Number : 1217852-49-7
  • Molecular Weight : 418.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/041799.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/041799.html
Availability details for
Quantity
  • Availability
  • Add to Cart