MAT236031757 Matrix Scientific

Fmoc-Cys(Dpm)-OH

  • MDL number MFCD02682541
  • Cas Number : 247595-29-5
  • Molecular Weight : 509.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-78
Country of Origin USA
Product Link http://www.matrixscientific.com/041812.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041812.html
Availability details for
Quantity
  • Availability
  • Add to Cart