MAT236031785 Matrix Scientific

Boc-(2S,3RS)-2-amino-3-hydroxy-4-methylpentanoicacid

  • MDL number MFCD02682586
  • Cas Number : 1217531-78-6
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/042068.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042068.html
Availability details for
Quantity
  • Availability
  • Add to Cart