MAT236031822 Matrix Scientific

Fmoc-6-methyl-DL-tryptophan

  • MDL number MFCD02682377
  • Cas Number : 1219150-44-3
  • Molecular Weight : 440.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/042153.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042153.html
Availability details for
Quantity
  • Availability
  • Add to Cart