MAT236031824 Matrix Scientific

Fmoc-7-methyl-DL-tryptophan

  • MDL number MFCD02682379
  • Cas Number : 138775-53-8
  • Molecular Weight : 440.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/042156.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042156.html
Availability details for
Quantity
  • Availability
  • Add to Cart